CO₂ incubators with hot air sterilisation and humidity regulation, CBF Series
Supplier: Binder
9640-0017 9640-0018 9640-0021 9640-0022 9640-0025
390-1618EA 12762 EUR
390-1618 390-1619 390-1622 390-1623 390-1626
CO₂ incubators with hot air sterilisation and humidity regulation, CBF Series
Incubators CO₂ Incubators
The CBF series CO₂ incubators are the premium series of CO₂ incubators with humidity control. Units are suitable for all sensitive incubation applications and ensure optimal cell growth in GMP environments. A CBF series CO₂ incubator can be used for complex cultivation experiments or individual environments with hypoxic conditions thanks to the comprehensive range of options and accessories. Units ensure consistent results including in cell-based assays with multiwell plates.
  • Contamination-safe humidification system without water pan
  • Tried-and-tested anti-