Cell culture plates, 384-well, Small Volume™, CELLSTAR®
Supplier: Greiner Bio-One
784080_32
392-0371EA 710 EUR
392-0371
Cell culture plates, 384-well, Small Volume™, CELLSTAR®
Microplates Cell Culture Plates
PS, flat bottom, round wells, TC treated. These 384-well Low Volume™ microplates offer the same standardised footprint and height as regular polystyrene plates. This enables a direct change from larger volumes into the small volume range or even a combination of both plates.
  • Raised rims on each well minimise cross contamination
  • Small volume allows reagent savings