Culture tubes, reusable, with screw cap, KIMAX®, KIMBLE®
Supplier: DWK Life Sciences
SP45066A-13100 SP45066A-16100 SP45066A-16125 SP45066A-16150 SP45066A-20125 SP45066A-20150 SP45066A-25150 SP45066A-25200 SP45066-38200
392-0373EA 475 EUR
392-0373 SCERSP45066A-16100 SCERSP45066A-16125 SCERSP45066A-16150 SCERSP45066A-20125 392-0374 392-0375 392-0376 392-0372
Culture tubes, reusable, with screw cap, KIMAX®, KIMBLE®
Tubes Culture Tubes
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
  • Very good chemical resistance
  • Very good temperature resistance
  • Uniform wall thickness
Certifications: Tubes with PTFE-faced rubber-lined caps are designed from ASTM Specification E982, Type VI, Class B requirements.
Ordering information: Cap is supplied unattached.