Culture media bottles, square, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
342040-0125 342040-0650 342040-1000 342044-0125 342044-0650 342044-1000 342040-0250 842040-0650 842040-1000 842040-0125
215-1168EA 0 EUR
215-1168 215-1847 215-1848 215-1849 215-1850 215-1851 392-0378 392-0381 392-0397 NALG842040-0125
Culture media bottles, square, sterile, Nalgene®
Bottles Cell Culture Bottles
PET, clear, with thread or complete with white HDPE screw caps.
  • Good chemical resistance
  • Temperature resistant from −40 to +70 °C
  • Breakproof, suitable for airfreight, leakproof closure
Light breakproof bottles with outstanding gas diffusion properties for storing and transporting liquid media, buffer solutions and sera. Bottles and caps are gamma-sterilised and pyrogen-free. They are non-cytotoxic. With textured graduations and inner sealing ring for use of tamper-evident caps.
Certifications: Complying with the requirements of USP, Class VI, USP 66More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement