Lithium metavanadate ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Lithium vanadium trioxide, Lithium vanadium oxide, Lithium vanadate

39358.18 39358.30
39358.18EA 105 EUR
39358.18 39358.30
Lithium metavanadate ≥99.9% (metals basis)
Lithium metavanadate

Purity: 99.9% CAS No.: 15060-59-0 Molecular Formula: LiVO3 Formula Weight: 105.88 Physical Form: Powder UN No.: 3285 MDL No.: MFCD00053997

Formula: LiO₃V
Melting Pt: 605 °C
Density: 2.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00053997
CAS Number: 15060-59-0
EINECS: 239-124-7
UN: 3285
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR