Thiazole orange 10 mM DMSO

Supplier: Biotium

Synonyms: 1-Methyl-4-[(3-methyl-2(3H)-benzothiazolylidene)methyl]quinolinium p-tosylate

40077
40077.EA 159 EUR
40077.
Thiazole orange 10 mM DMSO
Thiazole orange

Thiazole orange (TO) is commonly used in reticulocyte analysis to stain residual RNA of blood cells, to stain DNA in agarose gels and capillary electrophoresis. Thiazole Orange stains the yeast nucleus, and is also able to stain bacteria and mammalian cells.


  • λEx/λEm (with DNA) = 512/533 nm
  • Supplied at 10 mM in DMSO
  • Store at 4°C and protect from light


In live yeast, TO shows both nuclear concentration and cytoplasmic signal; after fixation the staining becomes completely cytoplasmic. In live mammalian cells, thiazole orange initially stains mitochondria and then redistributes to the nucleus and cytoplasm. Thiazole orange can also stain live bacteria (gram-positive and gram-negative).

Formula: C₂₆H₂₄N₂O₃S₂
MW: 476.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00192499
CAS Number: 107091-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR