Chromium(III) telluride ≥99.5% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

40206.06 40206.14
40206.06EA 72.6 EUR
40206.06 40206.14
Chromium(III) telluride ≥99.5% (metals basis)
Chromium (III) telluride

Purity: 99.5% CAS No.: 12053-39-3 Molecular Formula: Cr2Te3 Formula Weight: 486.79 Physical Form: -60+100 Mesh Powder Density: 7.0 Appearance: Gray-black Odour: Odourless Melting Point: ca 1300° MDL No.: MFCD00016040

Formula: Cr₂Te₃
Melting Pt: ∼1300 °C
Density: 7.0
Storage Temperature: Ambient
MDL Number: MFCD00016040
CAS Number: 12053-39-3
EINECS: 235-003-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR