Scandium ≥99.9% (metals basis excluding Ta), ingot (max. 8-12% Ta)

Supplier: Thermo Fisher Scientific


Packaged under argon.

40229.03 40229.06
40229.03EA 289 EUR
40229.03 40229.06
Scandium ≥99.9% (metals basis excluding Ta), ingot (max. 8-12% Ta)
Scandium

Purity: 99.9% CAS No.: 7440-20-2 Physical Form: Packaged under argon Appearance: Grey or silvery Odour: Odourless MDL No.: MFCD00016323

Formula: Sc
Boiling Pt: 2730 °C (1013 hPa)
Melting Pt: 1541 °C
Density: 2.989 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016323
CAS Number: 7440-20-2
EINECS: 231-129-2
Merck Index: 12,08537

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR