Strontium molybdenum oxide 99%

Supplier: Thermo Fisher Scientific

40238.09 40238.18 40238.30 40238.A1
40238.09EA 56.6 EUR
40238.09 40238.18 40238.30 40238.A1
Strontium molybdenum oxide 99%
Strontium molybdenum oxide

Purity: 99.9% CAS No.: 13470-04-7 Molecular Formula: SrMoO4 Formula Weight: 247.56 Physical Form: -200 Mesh Powder Density: 4.540 Melting Point: dec. Solubility: Insoluble in water. Soluble in acid MDL No.: MFCD00049550

Formula: SrMoO₄
Melting Pt: Dec.
Density: 4.540
Storage Temperature: Ambient
MDL Number: MFCD00049550
CAS Number: 13470-04-7
EINECS: 236-730-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR