Manganese(II) bromide tetrahydrate ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Manganous bromide tetrahydrate, Manganese dibromide tetrahydrate, Manganese(II) bromide hydrate

40245.14 40245.22 40245.36
40245.14EA 41.4 EUR
40245.14 40245.22 40245.36
Manganese(II) bromide tetrahydrate ≥98%
Manganese (II) bromide tetrahydrate

Purity: 98% CAS No.: 10031-20-6 Molecular Formula: MnBr2.xH2O Formula Weight: 214.76(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: Pink Odour: Odourless Melting Point: 64.3° dec. Solubility: Soluble in water, alcohol Merck Reference: 14,5725 MDL No.: MFCD00149791

Formula: Br₂Mn·4H₂O
Melting Pt: 64.3 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149791
CAS Number: 10031-20-6
EINECS: 236-591-9
Merck Index: 14,5725

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR