Ammonium iodide ≥99.995% (metals basis)

Supplier: Thermo Fisher Scientific
Warning

40255.06 40255.14 40255.22
40255.06EA 96.1 EUR
40255.06 40255.14 40255.22
Ammonium iodide ≥99.995% (metals basis)
Ammonium iodide

Purity: 99.995% CAS No.: 12027-06-4 Molecular Formula: H4IN Formula Weight: 144.94 Physical Form: -4 Mesh Density: 2.514 Refractive Index: 1.7031 Sensitivity: Light and Air Sensitive and Hygroscopic Appearance: White to pale yellow Odour: Odourless Melting Point: 551° subl. Merck Reference: 14,529 MDL No.: MFCD00011424

Formula: NH₄I
Melting Pt: 551 °C (subl.)
Density: 2.514
Storage Temperature: Ambient
MDL Number: MFCD00011424
CAS Number: 12027-06-4
EINECS: 234-717-7
Merck Index: 14,529

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR