Potassium hexachlororuthenate(IV) (metal basis) (≥25,3% Ru) ≥99.95%, Premion®

Supplier: Thermo Fisher Scientific
Warning

40399.03 40399.06
40399.03EA 88.6 EUR
40399.03 40399.06
Potassium hexachlororuthenate(IV) (metal basis) (≥25,3% Ru) ≥99.95%, Premion®
Potassium hexachlororuthenate(IV) (metal basis)

Purity: 99.95% CAS No.: 23013-82-3 Molecular Formula: K2RuCl6 Formula Weight: 391.99 Physical Form: Cystalline Powder MDL No.: MFCD00050159

Formula: K₂RuCl₆
Storage Temperature: Ambient
MDL Number: MFCD00050159
CAS Number: 23013-82-3
EINECS: 245-381-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR