Terbium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade

Supplier: Thermo Fisher Scientific
Danger

40573.NCEA 425 EUR
40573.NC
Terbium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Terbium (III) perchlorate

CAS No.: 14014-09-6 Molecular Formula: Cl3O12Tb.6H2O Formula Weight: 565.37 (457.28anhy) Physical Form: Liquid Density: 1.5 Appearance: Colourless Odour: Odourless UN No.: 3211 MDL No.: MFCD00150362

Formula: Cl₃O₁₂Tb
MW: 457.277 g/mol
Density: 1.5
Storage Temperature: Ambient
MDL Number: MFCD00150362
CAS Number: 14014-09-6
EINECS: 237-826-8
UN: 3211
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR