Aluminium 99,99 Metals basis, wire, hard, Ø 0.025 mm (0.001 in)

Supplier: Thermo Fisher Scientific

Synonyms: Aluminum

40677.G1
40677.G1EA 272 EUR
40677.G1
Aluminium 99,99 Metals basis, wire, hard, Ø 0.025 mm (0.001 in)
Aluminium

Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈0.0013g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00134029

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR