Formaldehyde 37% (w/w) in aqueous solution stabilised ACS

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilization: Stabilised with methanol 10-15%

410730010 410731000 410730050
410730010.EA 45.7 EUR
410730010. 410731000. 410730050.
Formaldehyde 37% (w/w) in aqueous solution stabilised ACS
Formaldehyde
Formula: CH₂O
Melting Pt: –15 °C
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 1198
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear solution
Color scale ≤10 APHA
Stabilizer 10 to 15 % methanol
Residue after ignition ≤0.005 %
Titration 36.5 to 38.0 %
Chloride (Cl) ≤5 ppm
Sulfate (SO4) ≤0.002 %
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Titrable acid ≤0.006 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR