Titanium ≥99.995% (metals basis), slug, Length: 9.5 mm (0.37 in), Ø 9.5 mm (0.37 in)

Supplier: Thermo Fisher Scientific
Warning

41303.KJ 41303.KM
41303.KJEA 157 EUR
41303.KJ 41303.KM
Titanium ≥99.995% (metals basis), slug, Length: 9.5 mm (0.37 in), Ø 9.5 mm (0.37 in)
Titanium

Purity: 99.995% CAS No.: 7440-32-6 Physical Form: ≈3g/slug Odour: Odourless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR