Dichloromethane, anhydrous ≥99.7% stabilised

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methylene chloride

Stabilization: Stabilised with amylene
Packaged under argon in resealable ChemSeal™ bottles.

41835.AK 41835.K2 41835.M1
41835.AKEA 46 EUR
41835.AK 41835.K2 41835.M1
Dichloromethane, anhydrous ≥99.7% stabilised
Dichloromethane

Purity: 99.7+% CAS No.: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93 Physical Form: Liquid Density: 1.325 Refractive Index: 1.4244 Odour: Sweet Melting Point: -95° Boiling Point: 39-40° Beilstein Reference: 1730800 Merck Reference: 14,6063 UN No.: 1593 MDL No.: MFCD00000881


Solvent for anhydrous reactions, water <0.01%.

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39…40 °C
Melting Pt: –95 °C
Density: 1.325
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 14,6063
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR