Ethyl acetate 99.6% ACS, meetSpec_Reagent Ph.Eur.

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Acetic acid ethyl ester

423680010 423685000 423680025 423680250
423680010.EA 45 EUR
423680010. 423685000. 423680025. 423680250.
Ethyl acetate 99.6% ACS, meetSpec_Reagent Ph.Eur.
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 75…78 °C (760 mmHg)
Melting Pt: –83.5 °C
Density: 0.902 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.2 % (K.F.)
Density (20/20°C) 0.901 to 0.904
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.003 %
Substance darkened by H2SO4 passes test
Titrable acid ≤0.0009 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR