Germanium(IV) isopropoxide ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Warning

42673.03 42673.06 42673.14
42673.03EA 77.4 EUR
42673.03 42673.06 42673.14
Germanium(IV) isopropoxide ≥99.9% (metals basis)
Germanium (IV) isopropoxide

Purity: 99.9% CAS No.: 21154-48-3 Molecular Formula: C12H28GeO4 Formula Weight: 308.94 Flash Point: 68°(154°F) Physical Form: Liquid Density: 1.025 Refractive Index: 1.414 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Alcohol-like Boiling Point: 109°/30mm MDL No.: MFCD00015339

Formula: C₁₂H₂₈GeO₄
Boiling Pt: 109 °C (30 mmHg)
Density: 1.025
Flash Pt: 68 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015339
CAS Number: 21154-48-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR