Copper ≥99.9% (metals basis), spherical powder APS 10 µm

Supplier: Thermo Fisher Scientific
Warning

42689.18 42689.30
42689.18EA 49.3 EUR
42689.18 42689.30
Copper ≥99.9% (metals basis), spherical powder APS 10 µm
Copper

Purity: 99.9% CAS No.: 7440-50-8 Appearance: Reddish Odour: Odourless UN No.: 3089 MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1,III
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR