Ethyl formate, extra dry 98+%, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Formic acid ethyl ester

429241000
429241000.EA 35.1 EUR
429241000.
Ethyl formate, extra dry 98+%, AcroSeal®
Ethyl formate
Formula: C₃H₆O₂
Boiling Pt: 54 °C (760 mmHg)
Melting Pt: –80 °C
Density: 0.917 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003294
CAS Number: 109-94-4
UN: 1190
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.0 %
UV at 280 nm A: ≤0.015
Residue after evaporation ≤0.0003 %
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR