N,N-Diethylaniline borane 97%, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Borane - N,N-Diethylaniline Complex

430931000
430931000.EA 129 EUR
430931000.
N,N-Diethylaniline borane 97%, AcroSeal®
N,N-Diethylaniline borane
Formula: C₁₀H₁₈BN
MW: 163.07 g/mol
Melting Pt: 30…27 °C
Density: 0.92 g/cm³
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00013187
CAS Number: 13289-97-9
UN: 3148
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5220 to 1.5250 (20°C, 589 nm)
Titration ≥96 % (Active hydrogen)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR