Gold ≥99.9% (metals basis), ultrathin foil, Thickness: 1.0 µm

Supplier: Thermo Fisher Scientific

43149.FF
43149.FFEA 401 EUR
43149.FF
Gold ≥99.9% (metals basis), ultrathin foil, Thickness: 1.0 µm
Gold

Purity: 99.9% CAS No.: 7440-57-5 Odour: Odourless MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR