Cyanide, standard solution, Specpure®

Supplier: Thermo Fisher Scientific

43804.AE 43804.AP
43804.AEEA 235 EUR
43804.AE 43804.AP
Cyanide, standard solution, Specpure®
Standards Anionic Standards Cyanide Standards

Molecular Formula: KCN in 0,1% NaOH
Physical Form: Liquid
MDL No.: MFCD00468042


Cyanide, standard solution is used to dissolve metals and their ores. It is also used in clinical chemistry and in waste water treatment facilities to determine the concentration of dissolved and particulate materials.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR