Gold ≥99.9% (metals basis), spherical powder -200 mesh

Supplier: Thermo Fisher Scientific

43901.06 43901.03 43901.14
43901.06EA 1530 EUR
43901.06 43901.03 43901.14
Gold ≥99.9% (metals basis), spherical powder -200 mesh
Gold

Purity: 99.9% CAS No.: 7440-57-5 MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR