Mechanical overhead stirrer, RW 47 digital
Supplier: IKA
4050000
441-0316EA 7007 EUR
441-0316
Mechanical overhead stirrer, RW 47 digital
Stirrers Overhead Stirrers
Extremely powerful, mechanically controlled laboratory stirrer designed for highly viscous applications for quantities up to 200 L. Unit has a robust, ergonomic design with constant power drive, suitable for intensive mixing for use in laboratories and pilot plants. The stirrer is equipped with protection to avoid overheating of the motor by means of self-locking temperature limiter. Additionally, to secure bowls, an optional stirring shaft protector and clamping fixture are available.
  • Digital speed display
  • Infinitely adjustable speed in two speed ranges