VWR® Magnetic Stirrers, Boxed Sets
Supplier: VWR Collection
442-0513EA 83.1 EUR
442-0513 442-0514 442-0515 442-0516 442-0517 442-0518 442-0519 442-0520 442-0521 442-0522
VWR® Magnetic Stirrers, Boxed Sets
Stirrers Magnetic Stirring Bars
Set Octahedral 1, PTFE covered, 12 octagonal magnetic stirring rods with centre ring, in practical, transparent box. Set includes two stirrers with each of following dimensions, Lר (mm): 13×8, 15×8, 25×10, 38×10, 51×10 and 64×10.
Set Cross-Shaped, PTFE covered, five cross-shaped magnetic stirring rods in practical, transparent box. Set includes one stirrer with each of following dimensions, Lר (mm): 10×10, 20×20, 25×25, 30×30 and 38×38.
Set Oval 1, PTFE covered, 12 oval magnetic stirring rods in practical, transparent box. Set includes two stirrers with each of following dimensions,More Product Information