Microplate shaker, PMS-1000i
Supplier: GRANT INSTRUMENTS
PMS-1000I
444-0041EA 571 EUR
444-0041
Microplate shaker, PMS-1000i
Shakers Microplate Shakers
Compact microplate shaker which provides reliable and regulated shaking for 96- and 384-well microplates with easy set up and digital timer. Variable speed across a wide range may be adjusted to suit the application: A low speed ensures that the well contents are not ejected, more vigorous agitation provides effective aeration across the small surface area of each of the wells. Simple screw attachments allow quick and easy fitting of any standard depth plates, holding the plates securely in place. Ideal for use in immunology, biotechnology, microbiology and pharmacology applications.