Orbital shakers, Rotamax 120 and Unimax series
Supplier: Heidolph Instruments GmbH & Co.KG
543-12310-00 544-41200-00 543-12319-00 543-12310-06 542-10020-09 544-41200-06 543-12319-06 542-10020-02 543-12310-02 543-12319-02 544-41200-02
444-1310EA 1871 EUR
444-1310 444-1116 444-0085 444-1234 444-1235 444-1233 444-0197 444-1472 444-1476 444-1477 444-1489
Orbital shakers, Rotamax 120 and Unimax series
Shakers Orbital Shakers
Orbital shakers with analogue control dials for vibration frequency. The two dimensional swirling action produced is ideal for cultivating microorganisms and cell or tissue cultures. Motion is continuously adjustable.
  • Continuous or timer operation; timer function with audible signal for unattended operation: Unimax 1010 up to 999 min process time, Rotamax 120 and Unimax 2010 up to 120 min
  • Unimax models have digital display; for speed (Unimax 2010), for speed and time (Unimax 1010)
  • Modular system; combine Unimax 1010 with Incubator 1000 with