Microplate shakers, MicroPlate Genie® and Multi-MicroPlate Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
SI-0403 SI-0404 SI-4003 SI-4004 SI-4003A SI-0403A SI-0404A SI-4004A SI-0402A
444-9155EA 560 EUR
444-9155 444-9156 SCIISI-4003 444-0941 SCIISI-4003A SCIISI-0403A 444-0940 444-0942 444-0875
Microplate shakers, MicroPlate Genie® and Multi-MicroPlate Genie®
Shakers Microplate Shakers
The high speed and small vortexing orbit of the mixers produces a true vortexing action so samples in each well of the microplate are mixed efficiently. Ideal for use in molecular biology applications and immunology laboratories.
  • Holds most types of skirted microplates, even 384 well formats
  • Thorough mixing regardless of sample viscosity
  • Timer function from 1 to 60 min (analogue models) or 1 to 99 min (digital models) or continuous mode (all)
  • Digital models can be programmed with 'ON' and 'OFF' pulsing time between 0,1 s to 99 min.
  • Multi-MicroPlate Genie®