Mini orbital shakers, KM 2 and KM 2 Akku
Supplier: Edmund Bühler GmbH
6135000 6113000
444-9111EA 2046 EUR
444-9111 444-9109
Mini orbital shakers, KM 2 and KM 2 Akku
Shakers Orbital Shakers
Durable orbital shakers, with digital control, available as standard or battery operated versions. The Akku model’s battery gives runtime of at least 40 hours. Different vessels can be accommodated by choosing from the range of optional attachments available.
  • Shakers have continuous or timer operation from 0 to 99 hours 59 min
  • The integrated control unit with LED display facilitates easy setting of the required shaking speed and runtime
  • Shakers can be used at 4 to 45 °C and 95% relative humidity (non-condensing), in an incubator or cold room
  • Low