Dual action shaker, KL 2
Supplier: Edmund Bühler GmbH
6115000
444-9120EA 1865 EUR
444-9120
Dual action shaker, KL 2
Shakers Reciprocating Shakers
Laboratory shaker which has functionality to change motion from orbital to reciprocating without using any tools. Sturdy, maintenance free shaker, suitable for long term use. Accepts 9×250 ml Erlenmeyer flasks or 3×250 ml separating funnels. Variety of different vessels can be accommodated by choosing from the range of optional attachments.
  • Motion can be changed from orbital to reciprocating
  • Continuous operation or timer operation from 0 to 120 min
  • Infinitely variable speed control
  • Can be used at 5 to 50 °C (85% relative humidity), in an incubator or cold