Gallium metal ≥99.999% (metals basis)

Supplier: Thermo Fisher Scientific
Warning

44418.22 44418.36
44418.22EA 690 EUR
44418.22 44418.36
Gallium metal ≥99.999% (metals basis)
Gallium

Packaged in poylethylene bottle.


Purity: 99.999% CAS No.: 7440-55-3 Physical Form: May be solid or liquid due to ambient temperature UN No.: 2803 MDL No.: MFCD00134045

Formula: Ga
MW: 69.72 g/mol
Boiling Pt: 2403
Melting Pt: 29
Density: 5.904 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8 III
Merck Index: 12,04364

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR