Sodium hexafluoroaluminate ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Trisodium hexafluoroaluminate, Sodium hexafluoroaluminate(III)

44478.14 44478.22 44478.36
44478.14EA 67 EUR
44478.14 44478.22 44478.36
Sodium hexafluoroaluminate ≥99.9% (metals basis)
Sodium hexafluoroaluminate

Purity: 99.9% CAS No.: 13775-53-6 Molecular Formula: Na3AlF6 Formula Weight: 209.95 Physical Form: Powder Density: 2.90 Appearance: White Odour: Odourless Melting Point: 1000° UN No.: 3260 MDL No.: MFCD00003507

Formula: AlF₆Na₃
Melting Pt: 1000 °C
Density: 2.90
Storage Temperature: Ambient
MDL Number: MFCD00003507
CAS Number: 13775-53-6
EINECS: 237-410-6
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR