Potassium-tert-butoxide 25% (w/w) 2 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Potassium-2-methylpropan-2-olate, Potassium tert-butylate, Potassium tert-butanolate, Potassium t-butoxide

445831000 445838000
445831000.EA 209 EUR
445831000. 445838000.
Potassium-tert-butoxide 25% (w/w) 2 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Potassium-tert-butoxide
Formula: C₄H₉KO
Density: 0.86 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00012162
CAS Number: 865-47-4
UN: 2920
ADR: 8,I

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid (May develop some turbidity or precipitate)
Strength 1.90 to 2.20 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR