Neodymium(III) acetate hydrate ≥99.9%

Supplier: Thermo Fisher Scientific

Synonyms: Neodymium(III) acetate xhydrate, Neodymium triacetate hydrate

44587.30
44587.30EA 358 EUR
44587.30
Neodymium(III) acetate hydrate ≥99.9%
Neodymium(III) acetate hydrate

Purity: 99.9% CAS No.: 334869-71-5 Molecular Formula: Nd(OOCCH3)3.xH2O Formula Weight: 321.38(anhy) Physical Form: Crystalline MDL No.: MFCD00013040

Formula: C₆H₁₁NdO₇
Storage Temperature: Ambient
MDL Number: MFCD00013040
CAS Number: 334869-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR