Titanium(IV) 2-ethylhexanoate ≥97%

Supplier: Thermo Fisher Scientific
Warning

44602.22 44602.36 44602.XK
44602.22EA 404 EUR
44602.22 44602.36 44602.XK
Titanium(IV) 2-ethylhexanoate ≥97%
Titanium(IV) 2-ethylhexanoate

Purity: 97% CAS No.: 3645-34-9 Molecular Formula: C32H60O8Ti Formula Weight: 623.75 Physical Form: Liquid Odour: Odourless MDL No.: MFCD05663781

Formula: C₃₂H₆₀O₈Ti
Storage Temperature: Ambient
MDL Number: MFCD05663781
CAS Number: 3645-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR