Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform (20,6% Pd)

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Tris(dibenzylideneacetone)dipalladium(0) chloroform adduct, Tris(dibenylideneacetone)dipalladium-chloroform, Tris(dibenzylideneacetone)dipalladium-chloroform adduct

44969.03 44969.06
44969.03EA 126 EUR
44969.03 44969.06
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform (20,6% Pd)
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform

CAS No.: 52522-40-4 Molecular Formula: C51H42O3Pd2.CHCl3 Formula Weight: 1035.11 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Air and Moisture Sensitive Melting Point: 131-135° MDL No.: MFCD00075479

Formula: C₅₁H₄₂O₃Pd₂·CHCl₃
Melting Pt: 131…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075479
CAS Number: 52522-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR