Molybdenum standard solution, 10 mg/L Mo in 5% nitric acid with hydrofluoric acid (max. 1%) (from Mo), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
45260.AEEA 89.6 EUR
45260.AE
Molybdenum standard solution, 10 mg/L Mo in 5% nitric acid with hydrofluoric acid (max. 1%) (from Mo), standard for ICP, Specpure®
Standards Cationic Standards Molybdenum Standards
Certifications: Traceable to NIST