VWR®, Europium Standard Solution, 1000 mg/L Eu in dilute Nitric Acid (from Eu2O3), Standard for ICP
Supplier: VWR Chemicals
455342TEA 156 EUR
455342T 455344V
VWR®, Europium Standard Solution, 1000 mg/L Eu in dilute Nitric Acid (from Eu2O3), Standard for ICP
Standards Cationic Standards Europium Standards
Eu₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.