VWR® ARISTAR®, Gadolinium Standard Solution, 1000 mg/L Gd in dilute Nitric Acid (de Gd2O3), Standard for ICP
Supplier: VWR Chemicals
455362AEA 133.2 EUR
455362A 455364C
VWR® ARISTAR®, Gadolinium Standard Solution, 1000 mg/L Gd in dilute Nitric Acid (de Gd2O3), Standard for ICP
Standards Cationic Standards Gadolinium Standards
Gd₂O₃ in HNO₃ (2 to 5%).
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.