VWR® ARISTAR®, Silver Standard Solution, 10000 mg/L Ag in dil. Nitric Acid (from AgNO₃), Standard for ICP
Supplier: VWR Chemicals
456032MEA 299 EUR
456032M
VWR® ARISTAR®, Silver Standard Solution, 10000 mg/L Ag in dil. Nitric Acid (from AgNO₃), Standard for ICP
Standards Cationic Standards Silver Standards
AgNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.