Connection stopper
Supplier: GERHARDT
12-0351
458-2357EA 146 EUR
458-2357
Connection stopper
Stoppers Rubber Stoppers
This connection stopper is made from viton and is suitable for distribution heads with connection pipe Ø 32,7 mm.