VWR® ARISTAR®, Bromide Standard Solution, 1000 mg/L Br- in Water (from NaBr) Standard for Ion Chromatography
Supplier: VWR Chemicals
458042WEA 116 EUR
458042W 458044B
VWR® ARISTAR®, Bromide Standard Solution, 1000 mg/L Br- in Water (from NaBr) Standard for Ion Chromatography
Standards Anionic Standards Bromide Standards
Br in H₂O.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.