ARISTAR®VWR® , Potassium Standard Solution, 200 mg/L K in dilute Nitric Acid (from K), Standard for Ion Chromatography
Supplier: VWR Chemicals
458302AEA 96 EUR
458302A
ARISTAR®VWR® , Potassium Standard Solution, 200 mg/L K in dilute Nitric Acid (from K), Standard for Ion Chromatography
Standards Cationic Standards Potassium Standards
K in H₂O tr.HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.