High temperature circulators, Forte HT series
Supplier: JULABO GmbH
9800031 9800062 9800063 9800035 9800065 9800066
461-0171EA 13092 EUR
461-0171 461-0172 461-0173 461-0188 461-0189 461-0190
High temperature circulators, Forte HT series
Circulating Thermostats Closed Circulator Systems Heating Circulators
High temperature circulators can be used for temperature applications up to 400 °C; units provide powerful heating capacities for shortest heat-up times and rapid compensation of endothermic reactions. Applications include use with reactors, autoclaves, distillation columns, mini plant installations, pilot plants and production facilities. Because of the closed circuit system oxidation is reduced and unpleasant odours and vapours do not escape, even at high temperatures.
  • Automatic heat-up ensures gentle heating
  • Integrated programmer with six 60-step