Ultra-low temperature refrigerated circulators
Supplier: JULABO GmbH
9352755 9352753 9352752 9352756 9352793 9352791 9352790 9312681 9162681 9352755N150 9352755N 9352753N150 9352753N 9352752N150 9352752N 9352756N150 9352795N 9352796N 9352756N 9352793N 9352791N150 9352791N 9352790N150 9352790N
461-0496EA 24500 EUR
461-0496 461-1102 461-0495 461-0497 461-0500 461-0499 461-0498 461-0462 461-0404 461-0467 461-0459 461-1104 461-1103 461-0466 461-0458 461-0158 461-0465 461-1106 461-0460 461-0156 461-1105 461-0464 461-0468 461-0157
Ultra-low temperature refrigerated circulators
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Ultra-low temperature refrigerated circulators for heating and cooling. The units are suitable for external temperature applications with simultaneous operation in the bath tank. The units provide particularly high heating and cooling capacities for rapid heating and cooling times even for large volume, external systems. FP models offer proportional cooling control to ensure energy saving performance and to reduce heat generation to the environment. Models with the designation W are water cooled. All models have handles and castors which allow easy transportation and drain ports forMore Product Information