Temperature calibration baths
Supplier: JULABO GmbH
9352628 9352627 9352514 9352508
461-0573EA 11365 EUR
461-0573 461-0572 461-0571 461-0570
Temperature calibration baths
Circulating Thermostats Open Circulator Systems Calibration Circulators
Calibration baths have been designed for applications in calibration laboratories such as ISO, DKD, NAMAS, UKAS and conform to the requirements according to DIN EN ISO 9001:2000. Units have a chamber at a constant temperature level and can be used for precise calibration of temperature sensors, measuring instruments, thermometers according to national and international standards. Refrigerated models are suitable for use at temperatures from −30 to +200 °C as well as use as calibration baths for temperatures from 50 to 300 °C. With the optional precision reference sensor it is possible toMore Product Information