Calibration thermostats, RE and Proline range
Supplier: LAUDA
L001917 L001918 L001919 L001920 L001921 L001922 L001923 L001924 L001925 L001926 L001927 L001928 L001929 L001930 L001931 L001932 L001933 L001934
461-1247EA 7450 EUR
461-1247 461-1248 LAUDL001919 LAUDL001920 LAUDL001921 LAUDL001922 461-1202 461-1203 461-1200 461-1201 LAUDL001927 LAUDL001928 LAUDL001929 LAUDL001930 LAUDL001931 LAUDL001932 LAUDL001933 LAUDL001934
Calibration thermostats, RE and Proline range
Circulating Thermostats Open Circulator Systems Calibration Circulators
Calibration thermostats have been designed for applications in calibration laboratories such as ISO, DKD, NAMAS, UKAS and conform to the requirements according to DIN EN ISO 9001:2000. Units offer excellent temperature stability and distribution over a wide temperature range in the test chamber. They have a height adjustable usable space and a separate pre-chamber. All models have an RS232 interface. Connection for PT100 reference sensor except RE 212 J.
  • Temperature uniformity of ±0,01 K
  • RE models have a vario pump with 5 output