Circulation thermostats, thermoelectric, LOOP
Supplier: LAUDA
L000027 L000580 L002014 L002015 L002016 L002017
461-1371EA 2540 EUR
461-1371 461-1372 LAUDL002014 LAUDL002015 LAUDL002016 LAUDL002017
Circulation thermostats, thermoelectric, LOOP
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
The LOOP product range includes two thermo-electric circulation thermostats for external temperature control.
  • Compact units which are not heavy
  • Refrigerant-free temperature control with Peltier technology
  • Power-regulated ventilator fan
  • 3-button controls with OLED display
  • Heating and cooling functionality via the same components
  • Voltage: 100 to 240 V, 50/60 Hz
  • RS232 interface as standard
Temperature control is by means of a current flow via a semiconductor relay. Both heating and cooling can be achieved simply by reversing the direction of current flowMore Product Information